Eye and Face Shower, Galvanized Pipe, Encon® Safety Products
Supplier: Encon Safety Products
01050234 01050230 01050216 01050207
45500-212EA 4675.74 CAD
45500-212 45500-216 45500-218 45500-220
Eye and Face Shower, Galvanized Pipe, Encon® Safety Products
First Aid Supplies Showers
This combination shower/face and eye wash with bowl is made with schedule 80 hot dip galvanized steel piping.
  • ¹/₂"CP bronze EFW valve
  • 1" CP bronze shower stay open valve with CP ball
  • Stainless steel valve stem
  • Stainless steel pull road assembly with actuation graphic
Eyewash fountain have two choices that is ABS plastic heads with float off covers and acetal heads with hinged covers.
Designed with varying hydraulic conditions from 30 to 70 psi (206.8 to 482.6 kPa).
Facewash fountain is offers with ABS plastic heads with wye and float off covers.
Order Now
SPECIFICATIONS
Environmentally Preferable