Anti-CD16 / CD32 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cyanine7) [clone: 2.4G2]

Supplier: Tonbo Biosciences
60-0161-U025 60-0161-U100
76040-306EA 102.7 CAD
76040-306 76040-308
Anti-CD16 / CD32 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cyanine7) [clone: 2.4G2]
Antibodies
The 2.4G2 antibody is specific for a common epitope found in the extracellular regions of mouse Fc-receptors Fc-gamma II (CD32) and Fc-gamma III (CD16). These receptors are expressed by B cells, monocytes, macrophages, NK cells, dendritic cells, and neutrophils.

Type: Primary
Antigen: CD16 / CD32
Clonality: Monoclonal
Clone: 2.4G2
Conjugation: PE (Phycoerythrin)-Cyanine7
Epitope:
Host: Rat
Isotype: Rat IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR