Anti-IgG/IgA/IgM Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Tonbo Biosciences
72-8112-M001
76040-280EA 206 CAD
76040-280
Anti-IgG/IgA/IgM Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat anti-Human IgG + IgA + IgM - Affinity Pure, HRP Conjugate

Type: Secondary
Antigen: IgG/IgA/IgM
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR