XJ Autolysis™ E. coli strains, Zymo Research
Supplier: Zymo Research
T3021 T3031 T3041 T3051 T5021 T5031 T5041 T5051
76020-118EA 407.61 CAD
76020-118 76020-120 76020-122 76020-124 76020-488 76020-490 76020-492 76020-494
XJ Autolysis™ E. coli strains, Zymo Research
Protein Expression Protein Expression Systems
Alternative E.coli strains for bacterial transformation and lysis.
  • Straightforward transformation procedure with up to 10^8 - 10^9 transformants/μg plasmid.
  • Simple, fast, and controlled autolysis of E. coli.
  • Available with DE3 lysogen for T7 promoter transcription.
XJ Autolysis™ E. coli strains are a new alternative for bacterial transformation and lysis. These strains are efficiently lysed following arabinose-induced expression of the bacteriophage λ endolysin protein, coupled to a single freeze-thaw cycle. The strains simplify protein expression and purification. They are also applicable for nucleic acid purification, and available with a DE3 lysogen encoding the T7 polymerase for expressing recombinant proteins driven by the T7 promoter.
Order Now
SPECIFICATIONS
Environmentally Preferable