Mix and Go!™ Competent Cells, Zymo Research
Supplier: Zymo Research
T3019 T3020 T3013 T3017 T3003 T3005 T3007 T3009 T3010 T3011
76020-114EA 240.25 CAD
76020-114 76020-116 76020-482 76020-484 76020-486 76020-662 76020-664 76020-666 76020-668 76020-670 76020-672
Mix and Go!™ Competent Cells, Zymo Research
Protein Expression Protein Expression Systems
The Mix and Go!™ Competent Cells are pre made, chemically competent cells for simple and highly efficient DNA transformation.
  • Mix and Go!™ transformation procedure with transformation efficiencies of 108 - 109 transformants/μg of plasmid DNA
  • Simply add DNA and then spread
  • DNA transformation in as little as 20 seconds
  • Uses: bacterial transformations, DNA cloning, blue-white screening