Proteinase K
Supplier: Zymo Research
D3001-2-20
D3001-2-5
76020-322EA
82
CAD
76020-322
76020-324
Proteinase K
Enzymes
Proteinase K is a stable serine protease with broad substrate specificity and will degrade many proteins in their native conformation, even in the presence of detergents (e.g., SDS).
- Specifications: Lyophilized enzyme provided with Proteinase K Storage Buffer.
- Enzyme Commission Number: (EC 3.4.21.64)
- Source: Engyodontium album pH and Temperature Range: 4.0 to 12.0 (8.0 is optimum), 25 to 65°C.
- Specific Activity: Over 30 units/mg protein
- Unit Definition: One unit (U) of enzyme will hydrolyze urea-denatured hemoglobin to produce 1.0 μmole of tyrosine per minute at pH 7.5 at 37°C.
The enzyme is frequently used in molecular biology applications to digest unwanted proteins such as nucleases from DNA and/or RNA preparations from microorganisms, cells, and plants.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...