X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 2% in DMF
Supplier: Zymo Research
|
Danger
|
X1001-25
X1001-5
76020-506EA
116.24
CAD
76020-506
76020-508
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 2% in DMF
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Sterile, ready to use X-Gal solution.
- Concentration: 2% w/v in DMF
- Storage: -20° C
Formula:
C₁₄H₁₅BrClNO₆ Flash Pt: 58 °C Storage Temperature: Freezer |
MDL Number:
MFCD00005666 CAS Number: 7240-90-6 UN: 2265 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...