Zymo-Spin™ VI Columns, Zymo Research
Supplier: Zymo Research
C1013-20 C1013-10
76004-150EA 126.51 CAD
76004-150 76005-182
Zymo-Spin™ VI Columns, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Spin columns for the purification of DNA.
  • Format: Spin column.
  • Binding Capacity: DNA- 500 µg.
  • Compatibility: centrifuge, vacuum manifold, luer-lok bottom assembly.
The versatile Zymo-Spin™ VI column can be used either in centrifuges or on-vacuum manifolds for the purification of DNA and/or RNA. Exclusive to this column is a luer-lok bottom assembly. The Zymo-Spin™ VI features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 500 μg DNA or RNA in ≥ 1 mL eluate. Capacity is 15 mL.