Zymo-Spin™ VE Columns, Zymo Research
Supplier: Zymo Research
C1021-25 C1024-25 C1024-50
76004-168EA 178.84 CAD
76004-168 76004-174 76004-200
Zymo-Spin™ VE Columns, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Spin columns for the purification of DNA or RNA.
  • Format: Spin column
  • DNA binding capacity: 125 µg
  • RNA binding capacity: 250 µg
  • Compatibility: microcentrifuge, centrifuge, vacuum manifold, syringe(luer-lok top)
The versatile Zymo-Spin™ V-E column can be used either in microcentrifuges, centrifuges, or on-vacuum manifolds for the purification of DNA and/or RNA. This column features a luer-lok top allowing it to be easily attached to a syringe, reservoir, or prefilter. The Zymo-Spin™ V-E features durable polypropylene construction and contains a unique silica-based matrix for the purification of up to 125 μg DNA or RNA in ≥ 100 μL elution buffer or water. The capacity of the spin column is 400 μL.
The Zymo-Spin™ V-E with Zymo Midi Filter™ assembly for purifying DNA/RNA from ≤15 mL sample.