Zymo-Spin™ V Columns, Zymo Research
Supplier: Zymo Research
Spin columns for the purification of DNA or RNA.
- Format: Spin column
- DNA binding capacity: 100 µg
- RNA binding capacity: 200 µg
- Compatibility: microcentrifuge, centrifuge, vacuum manifold, syringe(luer-lok top)
The versatile Zymo-Spin™ V column can be used either in microcentrifuges, centrifuges, or on-vacuum manifolds for the purification of DNA and/or RNA. This column features a luer-lok top allowing it to be easily attached to a syringe. The Zymo-Spin™ V features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 100 μg DNA or RNA in ≥ 100 μL eluate. Capacity is 800 μL.