Zymo-Spin™ III Columns, Zymo Research
Supplier: Zymo Research
C1005-250
C1005-50
76004-140EA
433.77
CAD
76004-140
76004-142
Zymo-Spin™ III Columns, Zymo Research
Nucleic Acid Reagents
Nucleic Acid Purification Kits and Reagents
Spin columns for the purification of DNA or RNA.
- Format: DNA/RNA binding
- DNA binding capacity: 25 µg
- RNA binding capacity: 100 µg
- Compatibility: microcentrifuge, vacuum manifold
The Zymo-Spin™ III column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ III features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 μg of DNA, and 100 μg of RNA, in ≥ 35 μL eluate. Capacity is 800 μL.