Zymo-Spin™ IIN Columns, Zymo Research
Supplier: Zymo Research
C1019-250
C1019-50
76004-162EA
433.77
CAD
76004-162
76004-164
Zymo-Spin™ IIN Columns, Zymo Research
Nucleic Acid Reagents
Nucleic Acid Purification Kits and Reagents
Spin columns for the purification of DNA or RNA.
- Format: DNA/RNA binding
- DNA binding capacity: 25 µg
- RNA binding capacity: 50 µg
- Compatibility: microcentrifuge, vacuum manifold
The Zymo-Spin™ IIN column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ IIN features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 μg of DNA, and 50μg of RNA, in ≥ 25 μL eluate. Capacity is 900 μL.