Zymo-Spin™ IIC Columns, Zymo Research
Supplier: Zymo Research
C1011-250 C1011-50
76005-174EA 576.86 CAD
76005-174 76005-176
Zymo-Spin™ IIC Columns, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Spin columns for the purification of DNA or RNA.
  • Format: DNA/RNA binding
  • DNA binding capacity: 25 µg
  • RNA binding capacity: 50 µg
  • Compatibility: microcentrifuge, vacuum manifold
The Zymo-Spin™ IIC column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ IIC features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 μg of DNA, and 50 μg of RNA, in ≥ 25 μL eluate. Capacity is 900 μL.