Zymo-Spin™ II Columns, Zymo Research
Supplier: Zymo Research
C1008-250
C1008-50
76005-164EA
433.77
CAD
76005-164
76005-166
Zymo-Spin™ II Columns, Zymo Research
Nucleic Acid Reagents
Nucleic Acid Purification Kits and Reagents
Spin columns for the purification of DNA or RNA.
- Format: DNA/RNA binding
- DNA binding capacity: 25 µg
- RNA binding capacity: 50 µg
- Compatibility: microcentrifuge
The Zymo-Spin™ II column features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 μg of DNA, and 50 μg of RNA, in ≥ 25 μL eluate. Capacity is 800 μL.