Zymo-Spin™ IC-XL Columns, Zymo Research
Supplier: Zymo Research
C1002-25 C1002-50
76005-144EA 67.61 CAD
76005-144 76005-146
Zymo-Spin™ IC-XL Columns, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Spin columns for the purification of DNA or RNA.
  • Format: DNA/RNA binding
  • DNA binding capacity: 10 µg
  • RNA binding capacity: 20 µg
  • Compatibility: microcentrifuge, vacuum manifold
The Zymo-Spin™ IC-XL column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ IC-XL features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 10 μg of DNA, and 20 μg of RNA, in ≥ 10 μL eluate. Capacity is 1 mL.