Zymo-Spin™ IB Columns, Zymo Research
Supplier: Zymo Research
C1014-250
C1014-50
76004-152EA
398.51
CAD
76004-152
76004-154
Zymo-Spin™ IB Columns, Zymo Research
Nucleic Acid Reagents
Nucleic Acid Purification Kits and Reagents
Spin columns for the purification of DNA and/or RNA and fluorescent dye removal.
- Format: DNA/RNA binding
- DNA binding capacity: 5 µg
- RNA binding capacity: 10 µg
- Compatibility: microcentrifuge, vacuum manifold
The black, opaque Zymo-Spin™ IB column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA and fluorescent dye removal. The Zymo-Spin™ IB features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 μg of DNA, and 10μg of RNA, in ≥ 6 μL eluate. Capacity is 800 μL.