Zymo-Spin™ I Columns, Zymo Research
Supplier: Zymo Research
C1003-250 C1003-50
76005-148EA 524.23 CAD
76005-148 76005-150
Zymo-Spin™ I Columns, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Spin columns for the purification of DNA or RNA.
  • Format: DNA/RNA binding
  • DNA binding capacity: 5 µg
  • RNA binding capacity: 10 µg
  • Compatibility: microcentrifuge, vacuum manifold
The Zymo-Spin™ I column can be used either in micro centrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ I features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 μg of DNA, and 10 μg of RNA, in ≥ 6 μL eluate. Capacity is 80 0μL.