RNA Clean & Concentrator™ Kits, Zymo Research

Supplier: Zymo Research

R1080 R1013 R1014 R1015 R1016 R1017 R1018 R1019
76020-414EA 1054.84 CAD
76020-414 76020-594 76020-604 76020-606 76020-608 76020-610 76020-612 76020-614
RNA Clean & Concentrator™ Kits, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

The RNA Clean & Concentrator™ (RCC™) kits facilitate the efficient removal of RNA polymerases, ligases, and RNA modifying enzymes.


  • Quick methods for cleaning and concentrating RNA
  • Zymo-Spin™ Column/Plate technology allows minimal elution volumes
  • Ideal for purification of RNA from aqueous phase following acid phenol extraction


The Zymo RNA Clean and Concentrator™ kits provide simple and reliable methods for the rapid preparation of high-quality RNA. The kit owes its simplicity to a unique single-buffer system and Zymo-Spin™ technology. Simply add the binding buffer to sample, adjust the conditions for binding by adding ethanol, then wash and elute the concentrated RNA. RNA ≥ 17 bases can be safely treated and recovered using these kits. The result is highly concentrated, purified RNA that is perfect for subsequent RNA-based methods, including RT-PCR, hybridization, and more.

Order Now

SPECIFICATIONS

Description RNA Clean & Concentrator™ RNA Clean & Concentrator™
Environmentally Preferable
Application
Target molecule RNA RNA
Refine target molecule Total RNA Total RNA
Sample type RNA preps RNA preps
Method / Format Multiwell plate Column (silica-based)
Prep size Mini Mini

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR