Mouse 5-mC and 5-hmC DNA Standards, Zymo Research
Supplier: Zymo Research
D5019 D5019-1 D5019-2 D5019-3 D5019-4
76020-076EA 824.16 CAD
76020-076 76020-078 76020-080 76020-082 76020-574
Mouse 5-mC and 5-hmC DNA Standards, Zymo Research
Standards Biological Standards
Control DNA for 5-mC and 5-hmC detection and/or quantitation applications.
  • Matched DNA set of genomic DNA from multiple organs
  • Precisely quantified levels of 5-mC and 5-hmC via LC/MS
  • Control DNA for 5-methylcytosine (5-mC) and 5-hydroxymethylcytosine (5-hmC) detection and/or quantitation applications (i.e. - mass spectrometry, HPLC, TLC, etc.)
Matched DNA Sets are an ideal control for detection and/or quantification methods against 5-methylcytosine (5-mC) and 5-hydroxymethylcytosine (5-hmC) as both modified cytosines are present at physiologically relevant levels and loci.
The Mouse 5-hmC & 5-mC DNA Set is a set of organ specific mouse genomic DNAs isolated from a pool of 8-10 week old Swiss Webster mice. The levels of 5-methylcytosine and 5-hydroxymethylcytosine have been precisely quantified by mass spectrometry (LC/MS).