DNA/RNA Shield™ Lysis Tubes, Zymo Research
Supplier: Zymo Research
R1104
R1105
R1102
R1103
76020-096EA
507.81
CAD
76020-096
76020-098
76020-424
76020-426
DNA/RNA Shield™ Lysis Tubes, Zymo Research
Buffers
Extraction Buffers
Collect and inactivate samples in lysis tubes prefilled with DNA/RNA Shield™.
- Nucleic acid preservation (at ambient temperature; cold-free)
- Pathogen inactivation (bacteria, fungus, parasites & viruses)
- Streamlined purification (no reagent removal, universally compatible, automatable)
Collect and inactivate samples in lysis tubes prefilled with DNA/RNA Shield™. Each tube is also filled with ultra-high density BashingBeads™, specifically designed for optimal tissue lysis. Samples collected are ready for any sensitive downstream analysis. Each lysis tube can also be paired with a sterile swab for initial sample handling.