DNA/RNA Shield™ Swab Collection Tubes, Zymo Research
Supplier: Zymo Research
R1106 R1107 R1108 R1109
76020-100EA 118.52 CAD
76020-100 76020-428 76020-430 76020-432
DNA/RNA Shield™ Swab Collection Tubes, Zymo Research
Swabs Swabs for Sampling
Swab collection system with tubes pre-filled with DNA/RNA Shield™ for collection and stabilization of nucleic acid at room termperature.
  • Nucleic acid preservation (at ambient temperature; cold-free)
  • Pathogen inactivation (bacteria, fungus, parasites & viruses)
  • Streamlined purification (no reagent removal, universally compatible, automatable)
A general swab collection system (12 x 80 mm screwcap tube) that allows for the collection of samples including mouth, nose, throat, etc
Applications:
Mouth, nose, and throat sample collection
Environmental sample collection
Pathogen inactivation and detection
The swab is collected into a tube prefilled with DNA/RNA Shield™, which effectively inactivates viral, bacterial, and other pathogens. Samples stored in DNA/RNA Shield™ are ready for downstream purification and any nucleic acid-based analysis.