Human Recombinant Cold-inducible RNA-binding protein (from E. coli)
Supplier: ADIPOGEN CORP MS
AG-40B-6000-C010 AG-40B-6000-3010
102980-354EA 343.08 CAD
102980-354 102980-356
Human Recombinant Cold-inducible RNA-binding protein (from E. coli)
Proteins and Peptides
Cold-inducible RNA-binding protein (CIRP) is from the family of cold shock proteins that plays a protective role in the genotoxic and cold stress response. CIRBP functions as an RNA chaperone to facilitate translation and to stabilize transcripts of genes involved in cell survival. CIRP (human) is a 172-aa nuclear protein consisting of one amino-terminal consensus sequence RNA-binding domain and one carboxyl-terminal glycine-rich domain. When overexpressed, CIRP promotes assembly of stress granules (SGs). CIRP is constitutively expressed at low levels in various tissues becoming up-regulated during mild hypothermia as well as exposure to UV irradiation and hypoxia. CIRP is also found extracellularly, where it acts as a proinflammatory mediator causing deleterious effects during hemorrhagic and septic shock.