Human Recombinant Neuregulin-4 (from E. coli)
Supplier: ADIPOGEN CORP MS
AG-40B-0155-C010 AG-40B-0155-3010
102980-338EA 438.46 CAD
102980-338 102980-340
Human Recombinant Neuregulin-4 (from E. coli)
Proteins and Peptides
Neuregulin-4 (Nrg4) belongs to a small family of EGF-like (EGFL) domain-containing proteins that are synthesized as transmembrane precursors and undergo proteolytic cleavage. The EGF-like domain (aa 5-46) of Nrg4 (aa 1-53) directly binds to the receptors ErbB3 and 4. Nrg4 is a cold induced adipokine, highly expressed in adipose tissues and enriched in brown fat. It is increased during brown adipocyte differentiation and reduced in rodent and human obesity. It promotes neurite outgrowth and protects against diet-induced insulin resistance and hepatic steatosis through attenuating hepatic lipogenic signaling. This hepatic effect of Nrg4 is mediated by ErbB3 and ErbB4 signaling that negatively regulates de novo lipogenesis mediated by LXR and SREBP1c. This effect of Nrg4 on fatty liver and insulin resistance could lead to the development of Nrg4 as an effective therapeutic biological for the treatment of NAFLD and type 2 diabetes. GST-Nrg4 (aa 1-53) recombinant protein has been shown to mimic the effect of endogenous secreted Nrg4 on liver lipogenesis.