Human Recombinant DLL3 (from HEK293 Cells)
Supplier: ADIPOGEN CORP MS
AG-40B-0151-C010 AG-40B-0151-3010
102980-322EA 343.08 CAD
102980-322 102980-324
Human Recombinant DLL3 (from HEK293 Cells)
Proteins and Peptides
DLL3 (Delta-like protein 3; Delta3) inhibits primary neurogenesis. It plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm. DLL3 binds and activates Notch-1. Defects in DLL3 are the cause of spondylocostal dysostosis type 1 (SCDO1).