Recombinant Flagellin (from E. coli)
Supplier: ADIPOGEN CORP MS
AG-40B-0125-C010 AG-40B-0125-3010
102980-218EA 343.08 CAD
102980-218 102980-220
Recombinant Flagellin (from E. coli)
Proteins and Peptides
Flagellin is the subunit protein which polymerizes to form the filaments of bacterial flagella. It activates the innate immune system through the receptor Toll-like Receptor 5 (TLR5) and the intracellular protein NLRC4 (NLR family CARD domain-containing protein 4).