Mouse Recombinant PEDF (from CHO Cells)

Supplier: ADIPOGEN CORP MS

AG-40B-0118-C010 AG-40B-0118-3010
102980-192EA 440 CAD
102980-192 102980-194
Mouse Recombinant PEDF (from CHO Cells)
Proteins and Peptides

Pigment epithelium-derived factor (PEDF) is a 47kDa secreted glycoprotein that belongs to the non-inhibitory serpin family group. PEDF is widely expressed in adult and fetal tissues, including brain, spinal cord, plasma, bone, prostate, pancreas, heart and lung. PEDF acts as an angiogenesis inhibitor with neurotrophic, immunomodulation and antitumor properties. It functions as anti-angiogenic agent by counterbalancing the proangiogenic effect of VEGF. PEDF is one of the most abundant proteins released by adipocytes and induces insulin resistance in adipocytes and human skeletal muscle cells. Recently, it has been reported that PEDF is sufficient to maintain the self-renewal of pluripotent human embryonic stem cells.

Order Now

SPECIFICATIONS

Protein/peptide name PEDF
Environmentally Preferable
Protein synonyms Caspin|Serpin F1|Stromal Cell-derived Factor 3|Pigment Epithelium-derived Factor
Protein/peptide type Recombinant
Species Mouse
Source CHO cells
Conjugation Unconjugated
Gene ID P97298
Endotoxin level Low
Endotoxin content <0.01EU/µg
Purity 95%
Concentration 0.1mg/ml after reconstitution.
Formulation Lyophilized. Contains PBS.
Molecular weight ~47kDa (SDS-PAGE)
Storage conditions –20 °C
Shipping temperature -20 °C, Blue Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR