Human Recombinant RANKL (Soluble) (from HEK293 Cells)
Supplier: ADIPOGEN CORP MS
AG-40B-0008-C010 AG-40B-0008-3010
102980-108EA 324.62 CAD
102980-108 102980-110
Human Recombinant RANKL (Soluble) (from HEK293 Cells)
Proteins and Peptides
RANKL is an osteoclast differentiation and activation factor. Augments the ability of dendritic cells to stimulate naive T cell proliferation. Important regulator of interactions between T cells and dendritic cells and may play a role in the regulation of the T cell-dependent immune response. RANKL plays an important role in the progression of breast cancer.