SiliaSep™ Tosic Acid (SCX) Flash Cartridges, SiliCycle
Supplier: SiliCycle
FR60530BI1500 FR60530BIS120 FR60530BIS120B FR60530BIS220 FR60530BIS220B FR60530BIS330 FR60530BIS330B FR60530BIS750 FR60530BISO04 FR60530BISO04B FR60530BISO12 FR60530BISO12B FR60530BISO25 FR60530BISO25B FR60530BISO40 FR60530BISO40B FR60530BISO80 FR60530BISO80B FR60530BI1500 FR60530BIS120 FR60530BIS120B FR60530BIS220 FR60530BIS220B FR60530BIS330 FR60530BIS330B FR60530BIS750 FR60530BISO04 FR60530BISO04B FR60530BISO12 FR60530BISO12B FR60530BISO25 FR60530BISO25B FR60530BISO40 FR60530BISO40B FR60530BISO80 FR60530BISO80B
75880-078BX 22261.38 CAD
75880-078 75880-080 75880-082 75880-084 75880-086 75880-088 75880-090 75880-092 75880-094 75880-096 75880-098 75880-100 75880-102 75880-104 75880-106 75880-108 75880-110 75880-112 CA75880-078 CA75880-080 CA75880-082 CA75880-084 CA75880-086 CA75880-088 CA75880-090 CA75880-092 CA75880-094 CA75880-096 CA75880-098 CA75880-100 CA75880-102 CA75880-104 CA75880-106 CA75880-108 CA75880-110 CA75880-112
SiliaSep™ Tosic Acid (SCX) Flash Cartridges, SiliCycle
Chromatography Columns
With its SiliaSep™ line, SiliCycle offers pre-packed Flash cartridges of superior performance.
  • Highest silica gel quality
  • Innovative packing technology
  • Universal Luer fittings
  • Compatible with any flash system
  • Cost effective
  • Multiple sorbent variety and cartridge sizes
With a more tightly packed silica bed and a homogeneous packing, the use of pre-packed flash cartridges improves purification efficiency by offering superior reproducibility and productivity compared to conventional manual flash chromatography.
SiliaSep Flash cartridges offers outstanding performance and reliability for all your separation requirements, from milligrams to kilograms.