SiliaSep™ TMA Chloride (SAX) nec Flash Cartridges, SiliCycle
Supplier: SiliCycle
FR66530BISO12 FR66530BISO12B FR66530BISO25 FR66530BISO25B FR66530BI1500 FR66530BIS120 FR66530BIS120B FR66530BIS220 FR66530BIS220B FR66530BIS330 FR66530BIS330B FR66530BIS750 FR66530BISO04 FR66530BISO04B FR66530BISO40 FR66530BISO40B FR66530BISO80 FR66530BISO80B FR66530BISO12 FR66530BISO12B FR66530BISO25 FR66530BISO25B FR66530BI1500 FR66530BIS120 FR66530BIS120B FR66530BIS220 FR66530BIS220B FR66530BIS330 FR66530BIS330B FR66530BIS750 FR66530BISO04 FR66530BISO04B FR66530BISO40 FR66530BISO40B FR66530BISO80 FR66530BISO80B
75879-386BX 222.67 CAD
75879-386 75879-388 75879-390 75879-392 75880-150 75880-152 75880-154 75880-156 75880-158 75880-160 75880-162 75880-164 75880-166 75880-168 75880-170 75880-172 75880-174 75880-176 CA75879-386 CA75879-388 CA75879-390 CA75879-392 CA75880-150 CA75880-152 CA75880-154 CA75880-156 CA75880-158 CA75880-160 CA75880-162 CA75880-164 CA75880-166 CA75880-168 CA75880-170 CA75880-172 CA75880-174 CA75880-176
SiliaSep™ TMA Chloride (SAX) nec Flash Cartridges, SiliCycle
Chromatography Columns
With its SiliaSep™ line, SiliCycle offers pre-packed Flash cartridges of superior performance.
  • Highest silica gel quality
  • Innovative packing technology
  • Universal Luer fittings
  • Compatible with any flash system
  • Cost effective
  • Multiple sorbent variety and cartridge sizes
With a more tightly packed silica bed and a homogeneous packing, the use of pre-packed flash cartridges improves purification efficiency by offering superior reproducibility and productivity compared to conventional manual flash chromatography.
SiliaSep Flash cartridges offers outstanding performance and reliability for all your separation requirements, from milligrams to kilograms.