SiliaSep™ Thiol Flash Cartridges, SiliCycle
Supplier: SiliCycle
FR51030BI1500 FR51030BIS120 FR51030BIS220 FR51030BIS330 FR51030BIS750 FR51030BISO04 FR51030BISO12 FR51030BISO25 FR51030BISO40 FR51030BISO80 FR51030BI1500 FR51030BIS120 FR51030BIS220 FR51030BIS330 FR51030BIS750 FR51030BISO04 FR51030BISO12 FR51030BISO25 FR51030BISO40 FR51030BISO80
75879-986BX 17823.19 CAD
75879-986 75879-988 75879-990 75879-992 75879-994 75879-996 75879-998 75880-000 75880-002 75880-004 CA75879-986 CA75879-988 CA75879-990 CA75879-992 CA75879-994 CA75879-996 CA75879-998 CA75880-000 CA75880-002 CA75880-004
SiliaSep™ Thiol Flash Cartridges, SiliCycle
Chromatography Columns
With its SiliaSep™ line, SiliCycle offers pre-packed Flash cartridges of superior performance.
  • Highest silica gel quality
  • Innovative packing technology
  • Universal Luer fittings
  • Compatible with any flash system
  • Cost effective
  • Multiple sorbent variety and cartridge sizes
With a more tightly packed silica bed and a homogeneous packing, the use of pre-packed flash cartridges improves purification efficiency by offering superior reproducibility and productivity compared to conventional manual flash chromatography.
SiliaSep Flash cartridges offers outstanding performance and reliability for all your separation requirements, from miligrams to kilograms.