SiliaSep™ Silica Flash Cartridges, SiliCycle
Supplier: SiliCycle
FR10030BI1500 FR10030BISO40 FR10030BISO80 FR10030BIS120 FR10030BIS220 FR10030BIS330 FR10030BIS750 FR10030BISO04 FR10030BISO12 FR10030BISO25 FR10030BI1500 FR10030BISO40 FR10030BISO80 FR10030BIS120 FR10030BIS220 FR10030BIS330 FR10030BIS750 FR10030BISO04 FR10030BISO12 FR10030BISO25
75879-280BX 2362.24 CAD
75879-280 75879-882 75879-884 75880-412 75880-414 75880-416 75880-418 75880-420 75880-422 75880-424 CA75879-280 CA75879-882 CA75879-884 CA75880-412 CA75880-414 CA75880-416 CA75880-418 CA75880-420 CA75880-422 CA75880-424
SiliaSep™ Silica Flash Cartridges, SiliCycle
Chromatography Columns
With its SiliaSep™ line, SiliCycle offers pre-packed Flash cartridges of superior performance.
  • Highest silica gel quality
  • Innovative packing technology
  • Universal Luer fittings
  • Compatible with any flash system
  • Cost effective
  • Multiple sorbent variety and cartridge sizes
With a more tightly packed silica bed and a homogeneous packing, the use of pre-packed flash cartridges improves purification efficiency by offering superior reproducibility and productivity compared to conventional manual flash chromatography.
SiliaSep Flash cartridges offersMore Product Information