SiliaSep™ Pentafluorophenyl (PFP) Flash Cartridges, SiliCycle
Supplier: SiliCycle
FR67530BIS120 FR67530BIS220 FR67530BIS330 FR67530BISO04 FR67530BISO12 FR67530BISO25 FR67530BISO40 FR67530BISO80 FR67530BIS120 FR67530BIS220 FR67530BIS330 FR67530BISO04 FR67530BISO12 FR67530BISO25 FR67530BISO40 FR67530BISO80
75880-178BX 2875.33 CAD
75880-178 75880-180 75880-182 75880-184 75880-186 75880-188 75880-190 75880-192 CA75880-178 CA75880-180 CA75880-182 CA75880-184 CA75880-186 CA75880-188 CA75880-190 CA75880-192
SiliaSep™ Pentafluorophenyl (PFP) Flash Cartridges, SiliCycle
Chromatography Columns
With its SiliaSep™ line, SiliCycle offers pre-packed Flash cartridges of superior performance.
  • Highest silica gel quality
  • Innovative packing technology
  • Universal Luer fittings
  • Compatible with any flash system
  • Cost effective
  • Multiple sorbent variety and cartridge sizes
With a more tightly packed silica bed and a homogeneous packing, the use of pre-packed flash cartridges improves purification efficiency by offering superior reproducibility and productivity compared to conventional manual flash chromatography.
SiliaSep Flash cartridges offers outstanding performance and reliability for all your separation requirements, from milligrams to kilograms.