Phenylboronic acid ≥99%, crystalline powder

Supplier: CHEM-IMPEX INTERNATIONAL, INC

Synonyms: Phenylmetaboric acid, Dihydroxy(phenyl)borane, Phenyl boric acid, Benzeneboronic acid

26799-100G 26799-1KG 26799-25G
CH26799-100GEA 64.78 CAD
CH26799-100G CH26799-1KG CH26799-25G
Phenylboronic acid ≥99%, crystalline powder
Phenylboronic acid

Used in numerous cross coupling reactions to make carbon-carbon bond formation (Suzuki reaction). It is also used in biology schemes as receptors and sensors.


Synonyms: Dihydroxyphenylborane; Phenyl-boranediol.

Formula: C₆H₇BO₂
MW: 121.93 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00002103
CAS Number: 98-80-6
Merck Index: 13,01068

Order Now

Specification Test Results

Purity limit ≥99% (HPLC)
Appearance White to off-white crystalline powder
Solubility Soluble in methanol , sparingly soluble in water
Melting Point 215 - 220 °C
Infrared Spectrum Conforms to structure
Loss on Drying ≤ 0.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR