Pierce™ Fast Western Blotting Kit, SuperSignal™ West Femto Substrate, Thermo Scientific
Supplier: Thermo Scientific
35080 35081
CAPI35080EA 1098.47 CAD
CAPI35080 CAPI35081
Pierce™ Fast Western Blotting Kit, SuperSignal™ West Femto Substrate, Thermo Scientific
Blotting Kits
The Pierce™ Fast Western Blot Kit, SuperSignal™ West Femto Substrate contains optimised reagents that shorten the time to perform a typical Western blot from 4 hours to ~60 minutes
  • Fast assay with femtogram levels of detection in about 1 hour
  • No expensive hardware or vacuum required; no clogging issues
  • Optimised protocol makes Western blot analysis easier
  • Complete kits contain all components needed to block, probe and develop a blot
The Fast Western Blot Kit protocol is a quick and efficient method for developing Western blots without the need toMore Product Information