Brady® BMP®61 Series Printer Ribbon
Supplier: Brady
148135 148136 148128 148086 148128 148134 M61-R4310 148128 M61-R6710-WT M61-R6410 148129 M61-R6010
CA75840-073RL 44.73 CAD
75840-083 75840-072 75840-073 CA75840-073 75840-064 75840-081 76520-182 76520-196 75840-077 76520-184 75840-079 76520-186
Brady® BMP®61 Series Printer Ribbon
Printer Ribbons
BMP®61 Series ribbon cartridges allow users to change ribbons in seconds.
  • R4300 Series Ribbons offers strong environmental resistance against smearing and chemicals
  • R6000 Series Halogen-Free Resin formulation offers excellent solvent, scratch, and heat resistance
  • R6200 Series Ribbons offer superior scratch, smear, heat, and chemical resistance
  • R6400 Series Resin formulation offers superior chemical and abrasion resistance
  • R6600 Series Resin formulation offers excellent solvent and friction resistance
  • R6700 Series Resin/Wax formulation is a white ribbon that offers good chemical resistance
  • R6800 Series white resin ribbons offer excellent solvent, scratch, and smudge resistance
One-color ribbon for printing a single color onto the appropriate Brady® label material.