Adhesive Vinyl GHS Labels 'Diamond Vinyl Labels', Type B-7569, Brady
Supplier: Brady Worldwide
130769 130770 130766 130767 130768 130771 130772 130773 130774 130775 130776 130777
10810-238RL 263.52 CAD
10810-238 10810-240 10810-248 10810-250 10810-252 10810-268 10810-270 10810-272 10810-274 10810-276 10810-278 10810-280
Adhesive Vinyl GHS Labels 'Diamond Vinyl Labels', Type B-7569, Brady
Labels
Blank white labels pre-printed with red GHS diamonds on durable indoor/outdoor grade vinyl material.
  • Withstands exposure to moisture, chemicals, and handling
These GHS Diamond Labels are made for use with the Brady BBP®85 Sign and Label Printer. They are white labels pre-printed with red GHS diamonds, which allows the printer to create 2-color GHS/CLP chemical labels in one print pass.
Certifications: RoHS compliant.