SiliaSep™ Diol Flash Cartridges, SiliCycle
Supplier: SiliCycle
FR35030BISO25 FR35030BISO25B FR35030BISO40 FR35030BISO40B FR35030BISO80 FR35030BISO80B FR35030BI1500 FR35030BIS120 FR35030BIS120B FR35030BIS220 FR35030BIS220B FR35030BIS330 FR35030BIS330B FR35030BIS750 FR35030BISO04 FR35030BISO04B FR35030BISO12 FR35030BISO12B FR35030BISO25 FR35030BISO25B FR35030BISO40 FR35030BISO40B FR35030BISO80 FR35030BISO80B FR35030BI1500 FR35030BIS120 FR35030BIS120B FR35030BIS220 FR35030BIS220B FR35030BIS330 FR35030BIS330B FR35030BIS750 FR35030BISO04 FR35030BISO04B FR35030BISO12 FR35030BISO12B
75879-314BX 299.82 CAD
75879-314 75879-316 75879-318 75879-320 75879-322 75879-324 75879-946 75879-948 75879-950 75879-952 75879-954 75879-956 75879-958 75879-960 75879-962 75879-964 75879-966 75879-968 CA75879-314 CA75879-316 CA75879-318 CA75879-320 CA75879-322 CA75879-324 CA75879-946 CA75879-948 CA75879-950 CA75879-952 CA75879-954 CA75879-956 CA75879-958 CA75879-960 CA75879-962 CA75879-964 CA75879-966 CA75879-968
SiliaSep™ Diol Flash Cartridges, SiliCycle
Chromatography Columns
With its SiliaSep™ line, SiliCycle offers pre-packed Flash cartridges of superior performance.
  • Highest silica gel quality
  • Innovative packing technology
  • Universal Luer fittings
  • Compatible with any flash system
  • Cost effective
  • Multiple sorbent variety and cartridge sizes
With a more tightly packed silica bed and a homogeneous packing, the use of pre-packed flash cartridges improves purification efficiency by offering superior reproducibility and productivity compared to conventional manual flash chromatography.
SiliaSep Flash cartridges offers outstanding performance and reliability for all your separation requirements, from miligrams to kilograms.