SiliaSep™ Diamine Flash Cartridges, SiliCycle
Supplier: SiliCycle
FR49030BI1500 FR49030BIS120 FR49030BIS220 FR49030BIS330 FR49030BIS750 FR49030BISO04 FR49030BISO12 FR49030BISO25 FR49030BISO40 FR49030BISO80 FR49030BI1500 FR49030BIS120 FR49030BIS220 FR49030BIS330 FR49030BIS750 FR49030BISO04 FR49030BISO12 FR49030BISO25 FR49030BISO40 FR49030BISO80
75879-374BX 11713.22 CAD
75879-374 75879-376 75879-378 75879-380 75879-382 75879-384 75879-978 75879-980 75879-982 75879-984 CA75879-374 CA75879-376 CA75879-378 CA75879-380 CA75879-382 CA75879-384 CA75879-978 CA75879-980 CA75879-982 CA75879-984
SiliaSep™ Diamine Flash Cartridges, SiliCycle
Chromatography Columns
With its SiliaSep™ line, SiliCycle offers pre-packed Flash cartridges of superior performance.
  • Highest silica gel quality
  • Innovative packing technology
  • Universal Luer fittings
  • Compatible with any flash system
  • Cost effective
  • Multiple sorbent variety and cartridge sizes
With a more tightly packed silica bed and a homogeneous packing, the use of pre-packed flash cartridges improves purification efficiency by offering superior reproducibility and productivity compared to conventional manual flash chromatography.
SiliaSep Flash cartridges offersMore Product Information