SiliaSep™ Cyano (CN) Flash Cartridges, SiliCycle
Supplier: SiliCycle
FR38030BI1500 FR38030BIS120 FR38030BIS120B FR38030BIS220 FR38030BIS220B FR38030BIS330 FR38030BIS330B FR38030BIS750 FR38030BISO04 FR38030BISO04B FR38030BISO12 FR38030BISO12B FR38030BISO25 FR38030BISO25B FR38030BISO40 FR38030BISO40B FR38030BISO80 FR38030BISO80B FR38030BI1500 FR38030BIS120 FR38030BIS120B FR38030BIS220 FR38030BIS220B FR38030BIS330 FR38030BIS330B FR38030BIS750 FR38030BISO04 FR38030BISO04B FR38030BISO12 FR38030BISO12B FR38030BISO25 FR38030BISO25B FR38030BISO40 FR38030BISO40B FR38030BISO80 FR38030BISO80B
75879-326BX 9949.13 CAD
75879-326 75879-328 75879-330 75879-332 75879-334 75879-336 75879-338 75879-340 75879-342 75879-344 75879-346 75879-348 75879-350 75879-352 75879-354 75879-356 75879-358 75879-360 CA75879-326 CA75879-328 CA75879-330 CA75879-332 CA75879-334 CA75879-336 CA75879-338 CA75879-340 CA75879-342 CA75879-344 CA75879-346 CA75879-348 CA75879-350 CA75879-352 CA75879-354 CA75879-356 CA75879-358 CA75879-360
SiliaSep™ Cyano (CN) Flash Cartridges, SiliCycle
Chromatography Columns
With its SiliaSep™ line, SiliCycle offers pre-packed Flash cartridges of superior performance.
  • Highest silica gel quality
  • Innovative packing technology
  • Universal Luer fittings
  • Compatible with any flash system
  • Cost effective
  • Multiple sorbent variety and cartridge sizes
With a more tightly packed silica bed and a homogeneous packing, the use of pre-packed flash cartridges improves purification efficiency by offering superior reproducibility and productivity compared to conventional manual flash chromatography.
SiliaSep Flash cartridges offers outstanding performance and reliability for all your separation requirements, from miligrams to kilograms.