SiliaSep™ C8 Flash Cartridges, SiliCycle
Supplier: SiliCycle
FR30830BI1500 FR30830BIS120 FR30830BIS220 FR30830BIS330 FR30830BIS750 FR30830BISO04 FR30830BISO12 FR30830BISO25 FR30830BISO40 FR30830BISO80 FR30830BI1500 FR30830BIS120 FR30830BIS220 FR30830BIS330 FR30830BIS750 FR30830BISO04 FR30830BISO12 FR30830BISO25 FR30830BISO40 FR30830BISO80
75879-886BX 15953.02 CAD
75879-886 75879-888 75879-890 75879-892 75879-894 75879-896 75880-426 75880-428 75880-430 75880-432 CA75879-886 CA75879-888 CA75879-890 CA75879-892 CA75879-894 CA75879-896 CA75880-426 CA75880-428 CA75880-430 CA75880-432
SiliaSep™ C8 Flash Cartridges, SiliCycle
Chromatography Columns
With its SiliaSep™ line, SiliCycle offers pre-packed Flash cartridges of superior performance.
  • Highest silica gel quality
  • Innovative packing technology
  • Universal Luer fittings
  • Compatible with any flash system
  • Cost effective
  • Multiple sorbent variety and cartridge sizes
With a more tightly packed silica bed and a homogeneous packing, the use of pre-packed flash cartridges improves purification efficiency by offering superior reproducibility and productivity compared to conventional manual flash chromatography.
SiliaSep Flash cartridges offers outstanding performance and reliability for all your separation requirements, from miligrams to kilograms.