SiliaSep™ Amine (WAX) Flash Cartridges, SiliCycle
Supplier: SiliCycle
FR52030BIS220 FR52030BISO04B FR52030BI1500 FR52030BIS120B FR52030BIS220B FR52030BISO80 FR52030BISO40B FR52030BIS120 FR52030BISO80B FR52030BISO40 FR52030BISO25B FR52030BISO12 FR52030BIS120 FR52030BIS120B FR52030BISO80 FR52030BIS750 FR52030BIS330B FR52030BI1500 FR52030BISO04B FR52030BIS330 FR52030BIS750 FR52030BISO40 FR52030BISO25 FR52030BIS330 FR52030BISO04 FR52030BIS220B FR52030BISO12B FR52030BISO04 FR52030BISO12 FR52030BISO40B FR52030BIS330B FR52030BISO25 FR52030BISO25B FR52030BIS220 FR52030BISO12B FR52030BISO80B
CA75880-056BX 3803.99 CAD
CA75880-056 75880-048 CA75880-058 CA75880-070 75880-062 CA75880-050 CA75880-072 75880-042 75880-064 CA75880-074 CA75880-052 75880-044 75880-066 CA75880-076 CA75880-054 75880-046 75880-068 75880-060 75880-058 CA75880-046 CA75880-068 CA75880-048 75880-050 75880-072 CA75880-060 75880-074 75880-052 CA75880-062 75880-054 75880-076 CA75880-042 CA75880-064 75880-056 CA75880-044 CA75880-066 75880-070
SiliaSep™ Amine (WAX) Flash Cartridges, SiliCycle
Chromatography Columns
With its SiliaSep™ line, SiliCycle offers pre-packed Flash cartridges of superior performance.
  • Highest silica gel quality
  • Innovative packing technology
  • Universal Luer fittings
  • Compatible with any flash system
  • Cost effective
  • Multiple sorbent variety and cartridge sizes
With a more tightly packed silica bed and a homogeneous packing, the use of pre-packed flash cartridges improves purification efficiency by offering superior reproducibility and productivity compared to conventional manual flash chromatography.
SiliaSep Flash cartridges offers outstanding performance and reliability for all your separation requirements, from miligrams to kilograms.