SiliaSep™ Alumina Neutral Flash Cartridges, SiliCycle
Supplier: SiliCycle
FAUT0054IS120 FAUT0054IS220 FAUT0054IS330 FAUT0054ISO04 FAUT0054ISO12 FAUT0054ISO25 FAUT0054ISO40 FAUT0054ISO80 FAUT0054IS120 FAUT0054IS220 FAUT0054IS330 FAUT0054ISO04 FAUT0054ISO12 FAUT0054ISO25 FAUT0054ISO40 FAUT0054ISO80
75879-264BX 1968.82 CAD
75879-264 75879-266 75879-268 75879-270 75879-272 75879-274 75879-276 75879-278 CA75879-264 CA75879-266 CA75879-268 CA75879-270 CA75879-272 CA75879-274 CA75879-276 CA75879-278
SiliaSep™ Alumina Neutral Flash Cartridges, SiliCycle
Chromatography Columns
With its SiliaSep™ line, SiliCycle offers pre-packed Flash cartridges of superior performance.
  • Highest silica gel quality
  • Innovative packing technology
  • Universal Luer fittings
  • Compatible with any flash system
  • Cost effective
  • Multiple sorbent variety and cartridge sizes
With a more tightly packed silica bed and a homogeneous packing, the use of pre-packed flash cartridges improves purification efficiency by offering superior reproducibility and productivity compared to conventional manual flash chromatography.
SiliaSep Flash cartridges offersMore Product Information