Brady® Chemical, Biohazard, and Hazardous Material Signs: Lead Hazard Do Not Enter Work Area Unless Authorized, Brady Worldwide
Supplier: Brady Worldwide
126023 126020 126022 126019 126021 126018
10810-382EA 30.6 CAD
10810-382 10810-384 10810-594 10810-596 10810-598 10810-600
Brady® Chemical, Biohazard, and Hazardous Material Signs: Lead Hazard Do Not Enter Work Area Unless Authorized, Brady Worldwide
Signs
Indoor/outdoor safety signs from Brady® are available in a variety of materials, allowing users to choose the material and size that best fits any application.
  • Self-Sticking Polyester (B-302) is an indoor/outdoor, high performance, pressure sensitive sign that is surface printed polyester with a polyester overlaminate
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter, or peel for up to 25 years
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial, and institutional environments
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial, and institutional environments
Signs that caution against an electrical hazard or identify chemicals and hazardous material storage, chemical containment areas, and acids or solvents provide the warnings and information that keep facilities and workers safe.
Products in this node are very specific to a particular chemical or hazard. Chemical names are often used in the header or text.
D.O.T. Hazard Signs are included here.
Certifications: Globally Harmonized System of Classification and Labeling of Chemicals (GHS). NFPA 704. OSHA 1910.1200.