Brady® Chemical, Biohazard, and Hazardous Material Signs: Hazardous Chemicals, Brady Worldwide
Supplier: Brady Worldwide
131765 131770 131769 131766 131768 131767 131763 131764
10810-468EA 24.63 CAD
10810-468 10810-472 10810-474 10810-476 10810-512 10810-514 10810-684 10810-686
Brady® Chemical, Biohazard, and Hazardous Material Signs: Hazardous Chemicals, Brady Worldwide
Signs
Indoor/outdoor safety signs from Brady® are available in a variety of materials, allowing users to choose the material and size that best fits any application.
  • Self-Sticking Polyester (B-302) is an indoor/outdoor, high performance, pressure sensitive sign that is surface printed polyester with a polyester overlaminate
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter, or peel for up to 25 years
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial, and institutional environments
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial, and institutional environments
Signs that caution against an electrical hazard or identify chemicals and hazardous material storage, chemical containment areas, and acids or solvents provide the warnings and information that keep facilities and workers safe.
Products in this node are very specific to a particular chemical or hazard. Chemical names are often used in the header or text.
D.O.T. Hazard Signs are included here.
Certifications: Globally Harmonized System of Classification and Labeling of Chemicals (GHS). NFPA 704. OSHA 1910.1200.