Brady® Chemical, Biohazard, and Hazardous Material Signs: Flammable Gas, Brady Worldwide
Supplier: Brady Worldwide
131797 131798 131801 131799 131800 131792 131791 131802 131789 131794 131793 131790 131788 131787 131795 131796
10810-494EA 24.67 CAD
10810-494 10810-498 10810-500 10810-520 10810-522 10810-528 10810-530 10810-540 10810-542 10810-544 10810-546 10810-548 10810-698 10810-702 10810-708 10810-716
Brady® Chemical, Biohazard, and Hazardous Material Signs: Flammable Gas, Brady Worldwide
Signs
Indoor/outdoor safety signs from Brady® are available in a variety of materials, allowing users to choose the material and size that best fits any application.
  • Self-Sticking Polyester (B-302) is an indoor/outdoor, high performance, pressure sensitive sign that is surface printed polyester with a polyester overlaminate
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter, or peel for up to 25 years
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial, and institutional environments
  • Durable Non-
More Product Information