Brady® Chemical, Biohazard, and Hazardous Material Signs: Explosives, Brady Worldwide
Supplier: Brady Worldwide
131773 131774 131777 131778 131775 131776 131771 131772
10810-488EA 23.29 CAD
10810-488 10810-490 10810-492 10810-496 10810-516 10810-518 10810-704 10810-706
Brady® Chemical, Biohazard, and Hazardous Material Signs: Explosives, Brady Worldwide
Signs
Indoor/outdoor safety signs from Brady® are available in a variety of materials, allowing users to choose the material and size that best fits any application.
  • Self-Sticking Polyester (B-302) is an indoor/outdoor, high performance, pressure sensitive sign that is surface printed polyester with a polyester overlaminate
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter, or peel for up to 25 years
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial, and institutional environments
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial, and institutional environments
Signs that caution against an electrical hazard or identify chemicals and hazardous material storage, chemical containment areas, and acids or solvents provide the warnings and information that keep facilities and workers safe.
Products in this node are very specific to a particular chemical or hazard. Chemical names are often used in the header or text.
D.O.T Hazard Signs are included here.
Certifications: Globally Harmonized System of Classification and Labeling of Chemicals (GHS). NFPA 704. OSHA 1910.1200.