Brady® Chemical, Biohazard, and Hazardous Material Signs: Diesel Fuel, Brady Worldwide
Supplier: Brady Worldwide
84376 60563 60533 122823 25428 25429 84375 87763 70267 60504 43006 122407
10810-068EA 30.6 CAD
10810-068 10810-342 10810-454 10810-482 10810-484 10810-486 10810-524 10810-526 10810-536 10810-538 10810-676 10810-694 10810-696
Brady® Chemical, Biohazard, and Hazardous Material Signs: Diesel Fuel, Brady Worldwide
Signs
Indoor/outdoor safety signs from Brady® are available in a variety of materials, allowing users to choose the material and size that best fits any application.
  • Self-Sticking Polyester (B-302) is an indoor/outdoor, high performance, pressure sensitive sign that is surface printed polyester with a polyester overlaminate
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter, or peel for up to 25 years
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial, and institutional environments
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial, and institutional environments
Signs that caution against an electrical hazard or identify chemicals and hazardous material storage, chemical containment areas, and acids or solvents provide the warnings and information that keep facilities and workers safe.
Products in this node are very specific to a particular chemical or hazard. Chemical names are often used in the header or text.
D.O.T. Hazard Signs are included here.
Certifications: Globally Harmonized System of Classification and Labeling of Chemicals (GHS). NFPA 704. OSHA 1910.1200.