Brady® Chemical, Biohazard, and Hazardous Material Signs: Corrosive, Brady Worldwide
Supplier: Brady Worldwide
22324 22323 40887 40888 84368 122650 70258 122822
10810-244EA 34.11 CAD
10810-244 10810-400 10810-410 10810-412 10810-532 10810-534 10810-556 10810-564
Brady® Chemical, Biohazard, and Hazardous Material Signs: Corrosive, Brady Worldwide
Signs
Indoor/outdoor safety signs from Brady® are available in a variety of materials, allowing users to choose the material and size that best fits any application.
  • Self-Sticking Polyester (B-302) is an indoor/outdoor, high performance, pressure sensitive sign that is surface printed polyester with a polyester overlaminate
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter, or peel for up to 25 years
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial, and institutional environments
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial, and institutional environments
Signs that caution against an electrical hazard or identify chemicals and hazardous material storage, chemical containment areas, and acids or solvents provide the warnings and information that keep facilities and workers safe.
Products in this node are very specific to a particular chemical or hazard. Chemical names are often used in the header or text.
D.O.T. Hazard Signs are included here.
Certifications: Globally Harmonized System of Classification and Labeling of Chemicals (GHS). NFPA 704. OSHA 1910.1200.