Brady® Chemical, Biohazard, and Hazardous Material D.O.T. Vehicle Placards: Class 8, Brady Worldwide
Supplier: Brady Worldwide
63420 60387
10810-236EA 68.65 CAD
10810-236 10810-432
Brady® Chemical, Biohazard, and Hazardous Material D.O.T. Vehicle Placards: Class 8, Brady Worldwide
Signs
D.O.T
B-946 vinyl (SSV) placards may be applied directly to a vehicle, and they provide over five years of outdoor durability. All placards come with square corners and no holes. Custom placards are also available upon request.
vehicle placards help ensure that hazardous material shipments reach their destinations without costly delays or rejections. All placards have square corners, no holes, and no headers.
Certifications: ANSI Z53.1-1967. ANSI Z535-2006. OSHA 1910.145.
Order Now
SPECIFICATIONS
Environmentally Preferable