Brady® Chemical, Biohazard, and Hazardous Material Signs: Chemical Storage, Brady Worldwide
Supplier: Brady Worldwide
84351 84352 22311 122820 70240 22312 122406 40876
10810-366EA 24.63 CAD
10810-366 10810-368 10810-408 10810-558 10810-562 10810-574 10810-718 10810-726
Brady® Chemical, Biohazard, and Hazardous Material Signs: Chemical Storage, Brady Worldwide
Signs
Indoor/outdoor safety signs from Brady® are available in a variety of materials, allowing users to choose the material and size that best fits any application.
  • Self-Sticking Polyester (B-302) is an indoor/outdoor, high performance, pressure sensitive sign that is surface printed polyester with a polyester overlaminate
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter, or peel for up to 25 years
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial, and institutional environments
  • Durable Non-
More Product Information